Learn More
R&D Systems™ Mouse Mesothelin (NP_061345.1) VersaClone cDNA
Product Code. 18702177
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18702177 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18702177 Supplier R&D Systems™ Supplier No. RDC3306
  • ONLINE EXCLUSIVE PRICE
    7945.00 NOK
    7514.85 NOK / 10µg
    Save 430.15 NOK
    5% Off
 Restricted Item
Estimated Shipment: 07-10-2026
Specifications
Gene Mesothelin
Accession Number NP_061345.1
Gene ID (Entrez) 10232
Gene Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg