Learn More
R&D Systems™ Mouse Melatonin R1A/MT1/MTNR1A (NP_032665) VersaClone cDNA
Specifications
Specifications
| Gene | Melatonin R1A/MT1/MTNR1A |
| Accession Number | NP_032665 |
| Gene ID (Entrez) | 4543 |
| Gene Alias | Mel1a receptor, MEL-1 A-R, melatonin receptor 1 A, melatonin receptor type 1 A, MelatoninR1A, MT1, MTNR1A |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |