Learn More
Novus Biologicals™ Mouse Lung Whole Tissue Lysate (Adult Whole Normal)
Product Code. 18036745 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18036745 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18036745 Supplier Novus Biologicals™ Supplier No. NBP305286
  • ONLINE EXCLUSIVE PRICE
    2565.00 NOK
    2423.40 NOK / 500µg
    Save 141.60 NOK
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Specifications
Quantity 500 μg
Purity Multi-step
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Target Lung