missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse LIF DuoSet ELISA

Product Code. 30001776
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 15
This item is not returnable. View return policy

Product Code. 30001776

Brand: R&D Systems™ DY449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

For the development of sandwich ELISAs to measure natural and recombinant mouse Leukemia Inhibitory Factor (LIF)

Based on its helical structure, LIF (Leukemia Inhibitory Factor) is considered a member of the Interleukin-6 family of cytokines. Functionally, it has been implicated in a many physiological processes including development, hematopoiesis, bone metabolism, and inflammation. Some cell types known to express LIF include activated T cells, monocytes, astrocytes, osteoblasts, keratinocytes, regenerating skeletal muscle, mast cells, and fibroblasts. The activities of LIF are mediated through a high-affinity heterodimeric receptor complex consisting of two membrane glycoproteins: an alpha subunit (LIF R alpha, also known as LIF R beta and CD118) that binds LIF with low affinity and the 130 kDa (gp130) subunit that does not bind LIF by itself, but is required for high-affinity binding of LIF by the complex.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Species Mouse
Conjugate Biotin
Binding Target LIF
Assay Range 6.25 to 400 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 3976
Synonym CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.