Learn More
MP Biomedicals Monkey Diet, Pellet
Product Code. 11447151 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
Unit Size:
10kg
20kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447151 10 kg 10kg
11457151 20 kg 20kg
11467151 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11447151 Supplier MP Biomedicals Supplier No. 0296025010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Monkey diet containing casein, special salt mixture, starch and special vitamin mixture.
  • Nutritionally complete for rhesus and old world monkeys
  • Special banana flavoring can also be added
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Product Type Animal Diet
Disposable Single-use
Format Pellet
Quantity 10 kg
Sterility Non-sterile
Storage Requirements Room Temperature
Length (English) 1 ft.
Length (Metric) 0.3048 m
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.