Learn More
Memmert™ MobileALERT For Up to 4 Alarm Notifications
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO105
ICO150
ICO240
ICO50
Unit Size:
Each
Description
- Accessory
- Notification by SMS, temperature and CO2 alarm standard
- Additionally humidity alarm when equipped with option K7
- O2 alarm when equipped with option T6
Specifications
Specifications
| Description | Temperature and humidity, up to 4 alarms |
| For Use With (Equipment) | ICO150 |