Learn More
Miele Professional™ Mobile injector
Shop All Miele Products
Click to view available options
:
Dimensions (D x W X H): 496 x 492 x 250mm
Description
With 16 nozzles (E 352) 6 x 220 mm with clips (E 354)
Specifications
Specifications
| Material | Stainless Steel, Plastic |
| Product Type | Insert/Module for Narrow-necked Glassware |
| Color | Stainless Steel, Black |
| Holds | 16 Narrow-necked glassware |
| For Use With (Equipment) | Washer-disinfectors, laboratory |
| Depth (Metric) | 496 mm |
| Height (Metric) | 250 mm |
| Width (Metric) | 492 mm |