Learn More
Miele Professional™ Mobile Injector
Product Code. 10353312 Shop All Miele Products
Click to view available options
:
Includes: 37 Injector Nozzles
Product Code. 10353312
Brand: Miele Professional™ 3810190
Only null left
Add to basket
Add to basket
Mobile unit for the optimum loading of narrow necked glassware
  • Equipped with 37 injector nozzles and locking mechanisms
  • Equipped with connection for hot air drying
  • Bracket for ML/2 for automatic mobile unit recognition
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mobile Injector Unit 1/1
Model E 414
For Use With (Equipment) Narrow-Necked Glassware
Includes 37 Injector Nozzles
Quantity 1 Each