Description
- Suitable for 23 pipettes arranged diagonally
- 10 pipettes up to 560 mm and 13 pipettes up to 490 mm
- Distance between holders 20 or 26 mm
Specifications
Specifications
| Material | Stainless Steel, Plastic |
| Product Type | Insert/Module for Pipettes |
| Color | Stainless Steel, Black |
| Holds | 23 pipettes |
| For Use With (Equipment) | Washer-disinfectors, laboratory |
| Depth (Metric) | 496 mm |
| Height (Metric) | 300 mm |
| Width (Metric) | 492 mm |