missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP12 Mouse anti-Human, Unconjugated, Clone: 1C6B5, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | MMP12 |
|---|---|
| Clone | 1C6B5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
16849235
|
Proteintech
66930-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16839235
|
Proteintech
66930-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The MMP12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MMP12. This antibody recognizes human antigen. The MMP12 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| MMP12 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 4321 | |
| Primary |
| 1C6B5 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human | |
| IgG1 |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts