Learn More
MMP12 Mouse anti-Human, Unconjugated, Clone: 1C6B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66930-1-IG-150UL
This item is not returnable.
View return policy
Description
The MMP12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MMP12. This antibody recognizes human antigen. The MMP12 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MMP12 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 1C6B5 | |
| Unconjugated | |
| 150 μL | |
| 4321 | |
| IgG1 |