Learn More
MMP12 Mouse anti-Human, Unconjugated, Clone: 1C6B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66930-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16849235
  • 1415.13 NOK / 20µL
Estimated Shipment: 12-10-2023
Description
The MMP12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MMP12. This antibody recognizes human antigen. The MMP12 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MMP12
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1C6B5
Unconjugated
20 μL
4321
IgG1