Description
Add channels to your MINIPULS™ 3 or Evolution and allow even greater flexibility in your laboratory.
MINIPULS 3 Pump Heads
Choose from these MINIPULS 3 pump heads:
- R1—Single channel pump head
- R4—Four channel pump head
- R8—Eight channel pump head
- R2—Two channel pump head
- R2HF—Two channel high flow pump head
- R4HF—Four channel high flow pump head
MINIPULS Evolution Pump Heads
Choose from these MINIPULS Evolution pump heads:
- MF1—Pump head with one cartridge
- MF4—Pump head with four cartridges
- HF2—Pump head with two high flow cartridges
Specifications
Specifications
| Flow Rate | 0.2μL/min. to 70 mL/min. |
| No. of Channels | 4 |
| Quantity | 1 Pack |
| Type | Pump Head |