Learn More
Fisherbrand™ Midi Centrifuge
Product Code. 12972041 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Depth (Metric):
171 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
12972041 171 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12972041
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
Achieve max speed with minimum space. Fisherbrand™ Midi Centrifuge is ideal for protocols calling for fast spins with a higher RCF requirement. Ideal for micro-filter cell separations, PCR, clinical applications and HPLC protocols. Includes two rotors: standard 12-place rotor with lid and eight-place strip rotor for four 0.2mL tube strips or 32 single tubes.
  • 5 Year Warranty
  • Compact footprint and brush-less motor for quiet operation and low vibration
  • Select and set the digital speed display in RCF or RPM
  • Sample imbalance and tilt safety cut-out
  • Safety interlock lid prevents opening while rotor is turning
  • Back lit display showing time, speed and operational information on the same screen
  • Quick spin button for short spin requirements
  • UV resistant plastic, increased useful life under harsh light conditions
  • Push-button lid release
  • Manual override allows user to open lid in case of power failure
Specifications
Capacity 12 x 1.5/2 mL
Max. Speed 12,500 rpm
Max. RCF 9,800 x g
Includes Rotors, 12 x 0.2mL and 12 x 0.5mL tube adapters, universal double insulated power adapter with four interchangeable plugs
Electrical Requirements 100/240 V 50/60 Hz
Depth (Metric) 171 mm
Rotor Included Yes
Dimensions (HWD) 114 x 171 x 203 mm
For Use With (Application) For PCR, microfilter separation, clinical applications and HPLC protocols.
Display Type Digital
Product Type Midi Centrifuge