Description
- Mini Spin Columns are the ideal solution to treat small volumes in combination with Selector Resins.
- They fit into every microcentrifuge and are compatible with all standard 1.5 ml or 2.0 ml tubes.
- Mini Spin Columns are provided with inserted 10 μm filters, screw caps, and snap-off plugs
Specifications
Specifications
| Kit Contents | 10 columns |
| For Use With (Application) | Rapid Purification of Recombinant Proteins |
For Research Use Only.