Learn More
Fisherbrand™ Horizontal Wide Format Mini-Plus Gel Unit
Product Code. 11553352 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Gel Dimensions:
L x W: 102 x 144 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Dimensions Unit Size
11553352 L x W: 102 x 144 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11553352
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
Includes
Gel unit, wide format, Mini-Plus, 1 x gel casting tray with gates, 2 x 1.0mm 20 sample combs, power supply connectors and loading strips
For faster separation of multiple samples.
  • For routine and rapid electrophoresis
  • Features four comb positions for fast separation of samples
  • Removable gel casting tray
Specifications
Type Horizontal Wide Format Mini-Plus Gel Unit
Gel Dimensions L x W: 102 x 144 mm
Includes
Gel unit, wide format, Mini-Plus, 1 x gel casting tray with gates, 2 x 1.0mm 20 sample combs, power supply connectors and loading strips
Sample Size 80
Volume (Metric) Buffer 500 mL