Learn More
Miele Professional™ Miele Process Documentation
Product Code. 15440445 Shop All Miele Products
Click to view available options
:
Software; additional licence
Product Code. 15440445
Brand: Miele Professional™ 10019160
APS 101 Segosoft Miele Edition (additional licences) for connection with a second or additional appliance.
  • User-controlled release of process reports for all machines
  • Protection from manipulation thanks to digital signature
  • Future-oriented product with regard to regulatory requirements
  • Only process documentation software with a quality seal
  • ServiceCard guarantees operation compliant with medical product
TRUSTED_SUSTAINABILITY
Specifications
Description APS 100 Segosoft Miele Edition (additional license)
Product Type Process/Operating Documentation/Communication
For Use With (Equipment) Washer-disinfectors, laboratory