Learn More
Miele Professional™ Miele Process Documentation
Product Code. 15490455 Shop All Miele Products
Click to view available options
:
category 6; model: type 9
Product Code. 15490455
Brand: Miele Professional™ 7951500
This item requires a price update prior to being ordered.
Request a Price for this item here.
Crossover network cable for direct connection to PC, length 5 m (type 9).
  • Cable category 6
  • Color: Transparent silver
  • Used for the network connection of Melag sterilizers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Process/Operating Documentation/Communication
For Use With (Equipment) Washer-disinfectors, laboratory