Learn More
Tenak™ Microtiterplates Comfort Rack For Upright Freezers
Rack designed to hold microtiterplates in upright freezers.
Brand:  Tenak™ TE35524
Additional Details : Weight : 0.55000kg
 View more versions of this product
Product Code. 12467070
  • 5060.00 NOK / Each
Estimated Shipment: 23-07-2024
Description
  • Variety of shelf heights available
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Includes an alert preventing the shelves from sliding out
Specifications
Specifications
Microtiterplate Rack
Plates (96)
540 x 272 x 135 mm
16 x 6
Upright Freezers
16 mm