Learn More
Tenak™ Microtiterplates Comfort Rack For Upright Freezers
Rack designed to hold microtiterplates in upright freezers.
4195.00 NOK - 5750.00 NOK
Specifications
Product Type | Microtiterplate Rack |
---|---|
For Use With (Equipment) | Upright Freezers |
Product Code | Brand | Height (Metric) Shelf | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) Shelf | Holds | Price | Quantity & Availability | ||||
12407100
|
Tenak™
TE35582 |
25 mm | Plates (56) |
4195.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
12417100
|
Tenak™
TE35584 |
25 mm | Plates (70) |
4545.00 NOK
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
||||
12467070
|
Tenak™
TE35524 |
16 mm | Plates (96) |
5060.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
12467090
|
Tenak™
TE35572 |
16 mm | Plates (91) |
4790.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
12616365
|
Tenak™
TE35528 |
16 mm | Plates (138) |
5750.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
15221004
|
Tenak™
TE35539 |
25 mm | Plates (90) |
5200.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
15231004
|
Tenak™
TE35549 |
45 mm | Plates (48) |
4220.00 NOK
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
Description
- Variety of shelf heights available
- Racks can either be used as one plate per shelf or the plates can be stacked
- Includes an alert preventing the shelves from sliding out
Specifications
Microtiterplate Rack | |
Upright Freezers |