Learn More
Eppendorf™ Centrifuge 5430/5430 R
BIOPHARMAGREENER_CHOICE
Product Code. 15865063 Shop All Eppendorf Products
230 V, 50/60 Hz (EU), With Rotor FA-45-30-11 incl. rotor lid, Keypad, Centrifuge 5430, 1 set
Click to view available options
Electrical Requirements:
230 V, 50/60 Hz (EU)
230 V/50-60 Hz (EU)
Includes:
With Rotor A-2-MTP
With Rotor FA-45-30-11 incl. rotor lid
Without rotor
Controller Type:
Keypad
Rotary Knob
Model:
Centrifuge 5430
Centrifuge 5430 (EU-IVD)
Centrifuge 5430 R
Centrifuge 5430 R (EU-IVD)
Unit Size:
1 set
Each
Product Code. 15865063
Brand: Eppendorf™ 5427000210
The unique, cross-over Eppendorf Centrifuge 5430 and 5430 R combine the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, they can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
  • Max. rotor capacity: 48 × 1.5/2.0 mL, 6 × 50 mL, 2 × MTP
  • Max. speed: 30,130 × g (17,500 rpm)
  • Remarkable versatility with 12 different rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Automatic rotor recognition and imbalance detection for maximized operational safety
  • Eppendorf QuickLock™ – System for quickly opening and closing the rotor lid
  • Greener Choice Claims
    • Environmental benefits include:
      • Renewable Energy
      • Ozone-Safe or Safe for the Ozone Layer
      • Non-Toxic
      • Recycled Content
      • Recyclable
      • Free-of a substance that causes environmental harm
    TRUSTED_SUSTAINABILITY
    Specifications
    Capacity 30 x 2 mL
    Max. Speed 17,500 rpm
    Max. RCF 30,130 x g
    Refrigerated No
    Display Large, brightly lit LCD
    Height (English) Exterior 9.8 in.
    Height (Metric) Exterior 25 cm
    Noise Level <58 dB(A) with Rotor F-45-30-11
    Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition
    Timer 30 sec. to 99:59 hrs., with continuous run function
    Width (English) Exterior 11.2 in.
    Width (Metric) Exterior 34 cm
    Wattage 475 W
    Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    Includes With Rotor FA-45-30-11 incl. rotor lid
    Dimensions (L x W x H) Exterior 330 x 420 x 250 mm
    Electrical Requirements 230 V, 50/60 Hz (EU)
    Controller Type Keypad
    Ventilated Yes
    Rotor Included Yes
    Standards EN ISO 12100, EN 50581, EN 61010-1, EN 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
    Frequency 50/60 Hz
    Voltage 230 V
    Model Centrifuge 5430
    Product Title
    Eppendorf™ Centrifuge 5430/5430 R >