Learn More
Applied Biosystems™ MicroAmp™ Cap Installing Tool
Product Code. 10746660 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Tool
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10746660 1 Tool Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10746660 Supplier Applied Biosystems™ Supplier No. 4330015
Estimated Shipment: 28-12-2026
MicroAmp™ Cap Installing Tool
The MicroAmp™ Cap Installing Tool helps you install strips of MicroAmp™ caps onto MicroAmp™ reaction tubes in a tray⁄retainer set or 96-well reaction plate.
  • Available in handle style
  • Seal 8 or 12 cap strips
  • Recessed cup for pressing caps onto tubes
  • Convenient tool ensures proper sealing
    Press and Seal Caps
    The MicroAmp™ Cap Installing Tool enables you to ergonomically seal MicroAmp™ reaction tubes and plates. This tight seal will prevent well-to-well contamination and sample loss.
    Used With
    MicroAmp™ Optical 8-Cap Strip, MicroAmp™ Optical 8-Tube Strip, 0.2 ml, MicroAmp™ Optical 96-Well Reaction Plate with Barcode, MicroAmp™ Optical 96-Well Reaction Plate with Barcode.
    Note: For Research Use Only. Not for use in diagnostics procedures.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Content And Storage Cap installing tool for MicroAmp plates, tubes, and caps
    Format 12-cap Strip, 8-cap Strip
    For Use With (Equipment) 2720 Thermal Cycler, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, GeneAmp 9700, QuantStudio™, SimpliAmp™ Thermal Cycler, StepOne™, StepOnePlus™, Veriti Dx Fast Thermal Cycler, Veriti Dx Thermal Cycler, Veriti Fast Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA™ 7 System, ProFlex™ PCR System, ViiA™ 7 Dx Fast System, 7500 Fast Dx System, 2720 Thermal Cycler, ViiA™ 7 Fast System, QuantStudio™ Dx
    For Use With (Application) Sequencing
    Product Line MicroAmp
    Product Type Cap Installing Tool
    Quantity 1 Tool
    Shipping Condition Room Temperature