Learn More
Fisherbrand™ Micro-Insert, Clear Glass, for small opening vial
Product Code. 11861653 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
0.2 mL
0.25 mL
0.3 mL
Height (Metric):
27.5 mm
29 mm
31 mm
Volume (Metric) Residual:
<1 μL
<2 μL
<8 μL
1 μL
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Height (Metric) Volume (Metric) Residual Unit Size
11861653 0.25 mL 31 mm <2 μL Pack of 1000
11762428 0.2 mL 31 mm <1 μL Pack of 1000
11772428 0.3 mL 31 mm <8 μL Pack of 1000
11858951 0.2 mL 29 mm <1 μL Pack of 1000
11848951 0.2 mL 27.5 mm 1 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11861653
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
Small opening vials require Inserts with 5mm diameter.
Small opening vials require Inserts with 5mm diameter.
  • Micro-Insert
  • For use with Small Opening Vials
  • Clear
  • Glass
TRUSTED_SUSTAINABILITY
Specifications
Description Micro Insert
Color Clear
Diameter (Metric) Outer 5 mm
Material Glass
Volume (Metric) 0.25 mL
For Use With (Application) Micro Sampling
Volume (Metric) Residual <2 μL
Base Style Conical Bottom
Quantity 10 x 100
Height (Metric) 31 mm