Learn More
Gilson™ MF1 Pump Head for Gilson™ Peristaltic Pump
Product Code. 12396112 Shop All Gilson Products
Change view
Click to view available options
Flow Rate:
0.2μL/min. to 70 mL/min.
Unit Size:
Each
Product Code. Flow Rate unitSize
12396112 0.2μL/min. to 70 mL/min. Each
1 options
Product Code. 12396112
Brand: Gilson™ F110705
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Add channels to your MINIPULS™ 3 or Evolution and allow even greater flexibility in your laboratory.
MINIPULS 3 Pump Heads
Choose from these MINIPULS 3 pump heads:
  • R1—Single channel pump head
  • R4—Four channel pump head
  • R8—Eight channel pump head
  • R2—Two channel pump head
  • R2HF—Two channel high flow pump head
  • R4HF—Four channel high flow pump head
MINIPULS Evolution Pump Heads
Choose from these MINIPULS Evolution pump heads:
  • MF1—Pump head with one cartridge
  • MF4—Pump head with four cartridges
  • HF2—Pump head with two high flow cartridges
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 0.2μL/min. to 70 mL/min.