Learn More
Mettler Toledo™ SevenGo™ perfectION Combined ISE Electrodes
Shop All Mettler Toledo Products
Click to view available options
Parameter:
Calcium
Chloride
Cyanide
Iodide
Nitrate
Silver/Sulfide
Unit Size:
Each
Description
- As vast as the range of different ions, is the selection of ionÂselective electrodes (ISE)
- The special Click & Clear™ junction allows for an optimal contact of the electrolyte solution and the sample
- With the dedicated solutions the sample can be optimally prepared for successful measurement of ion concentration
Specifications
Specifications
| Applications | Ag+/S-2 ions |
| Connector Types | Lemo |
| Filling Solution Type | ISA |
| Material | Epoxy |
| Operating Range (Metric) | 0.003 to 32000 mg/L |
| Temperature (English) Operating | 32° to 176°F |
| Electrode | Combined ISE Electrode |
| For Use With (Equipment) | All pH, mV Meters or Ionometers |
| Temperature (Metric) Operating | 0°C to 80°C |
| Type | Combination ISE |
| Show More |