Learn More
METTLER TOLEDO™ SevenExcellence™ S400 Benchtop pH Meter
Product Code. 13420679 Shop All Mettler Toledo Products
Change view
Click to view available options
Model:
Meter, Bio/Life Sciences Kit
Meter, Standard Kit
S400 pH Meter
Unit Size:
Each
Product Code. Model unitSize
13420679 Meter, Bio/Life Sciences Kit Each
13457806 Meter, Standard Kit Each
13440679 S400 pH Meter Each
3 options
Product Code. 13420679
Brand: METTLER TOLEDO™ 30046242
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The S400 meter measures pH, redox/ORP, and ISFET. It can be expanded with additional modules to measure conductivity, resistivity, DO, BOD, ISE, salinity, and TDS. It has an intuitive touchscreen, the ability to create measurement shortcuts, and can export data to a printer, Excel, or LIMS system.
  • pH, Redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, ISE, salinity, TDS, and BOD
  • Easily create measurement and calibration shortcuts
  • 7 inch color touch screen with intuitive menu navigation
  • Compatible with a wide range of electrodes for any application
  • *Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Export data to a printer, Excel file, or LIMS system
  • Automation available for measurements and cleaning in high-throughput applications
  • Manage user permissions and access to ensure data integrity
  • Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
  • Optional comprehensive qualification package available including IQ/OQ/PQ
  • Preventative maintenance and service available through METTLER TOLEDO Service
TRUSTED_SUSTAINABILITY
Specifications
Includes S400 pH meter; InLab Routine Pro-ISM electrode; electrode holder; ; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate; pH measurement guide; Multipin-BNC/RCA electrode cable 1.2 m
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.1
Accuracy (pH) ±0.002
Outputs USB, RS-232, Ethernet
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Calibration Points (pH) Max. 5 points, 8 pre-defined and 20 user-defined buffer group
Range (mV) -2000 to 2000
Resolution (mV) 0.1/1mV
Temperature Range (Metric) -30°C to +130°C
Resolution (Temperature) 0.1°C
Display Type TFT
Memory 250 analyses (20.000 data points)
Data Points 20,000
Model Meter, Bio/Life Sciences Kit
Range (Conductivity) 0.001μS/cm to 2000mS/cm
Languages Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Resolution (Conductivity) 0.001 to 1
Channels 1
Calibration Points (Conductivity) 13 pre-defined and 20 user-defined standard