Learn More
Mettler Toledo™ Electrode Fill Solutions
Product Code. 11917781 Shop All Mettler Toledo Products
Change view
Click to view available options
Filling Solution Type:
1M KCl; 25 mL
FRISCOLYT-B
KCl saturated with AgCl; 25mL
For Use With (Equipment):
DX Series ISE Half Cell
Reference Electrode
Unit Size:
25mL
Product Code. Filling Solution Type For Use With (Equipment) unitSize
11917781 1M KCl; 25 mL DX Series ISE Half Cell 25mL
12747453 FRISCOLYT-B Reference Electrode 25mL
10587335 KCl saturated with AgCl; 25mL Reference Electrode 25mL
3 options
Product Code. 11917781
Brand: Mettler Toledo™ 51343181
Made from the highest quality reagents to provide optimum performance.
  • Aqueous solution containing inorganic and organic components
  • Supplied in 25mL (0.9 oz.) bottle
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.9 oz.
Capacity (Metric) 25 mL
Concentration 1 mol/L
For Use With (Equipment) DX Series ISE Half Cell
Filling Solution Type 1M KCl; 25 mL