Learn More
(Methylthio)acetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Product Code. 11445297
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445297 25 g 25g
11435297 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11445297 Supplier Thermo Scientific Alfa Aesar Supplier No. B22667.14
 Restricted Item
In Stock
Add to basket
CAS: 40015-15-4 | Molecular Formula: C5H12O2S | Molecular Weight: 136.209 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40015-15-4
Molecular Formula C5H12O2S
Molecular Weight (g/mol) 136.209
MDL Number MFCD00009848
InChI Key DVOAQLUDKIFSNB-UHFFFAOYSA-N
Synonym methylthio acetaldehyde dimethyl acetal, 1,1-dimethoxy-2-methylthio ethane, ethane, 1,1-dimethoxy-2-methylthio, 1,1-dimethoxy-2-methylsulfanyl-ethane, ethane,1,1-dimethoxy-2-methylthio, methylmercapto acetaldehyde dimethyl acetal, 1,1-dimethoxy-2-methylsulfanyl ethane, acmc-209j9v, methylthio acetaldehyde dimethylacetal, 2-methylthio acetaldehyde dimethyl acetal
PubChem CID 638113
IUPAC Name 1,1-dimethoxy-2-methylsulfanylethane
SMILES COC(CSC)OC
Specifications
Density 1.022
Boiling Point 170°C to 171°C
Flash Point 53°C (127°F)
Refractive Index 1.4528
Quantity 25 g
UN Number UN1993
Beilstein 773689
Formula Weight 136.21
Percent Purity 97%
Chemical Name or Material (Methylthio)acetaldehyde dimethyl acetal