accessibility menu, dialog, popup

UserName

CH3OSH3COCH3

CAS RN 40015-15-4

IUPAC Name: 1,1-dimethoxy-2-(methylsulfanyl)ethane
Synonyms: 1,1-dimethoxy-2-(methylsulfanyl)ethane
Molecular Weight (g/mol): 136.21
Molecular Formula: C5H12O2S
InChi Key: DVOAQLUDKIFSNB-UHFFFAOYSA-N
SMILES: COC(CSC)OC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)