Learn More
MedChemExpress Methyl protodioscin
Product Code. 16491548
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491548 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16491548 Supplier MedChemExpress Supplier No. HYN0863
 Restricted Item
Estimated Shipment: 16-11-2026
MedChemExpress Methyl protodioscin(NSC-698790) is a furostanol bisglycoside with antitumor properties; shows to reduce proliferation, cause cell cycle arrest.
Specifications
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 1063.23 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 54522-52-0
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 140 mg/mL (131.67 mM∣Need ultrasonic) ∣H2O : 25 mg/mL (23.51 mM∣Need ultrasonic)
Synonym NSC-698790∣ Smilax saponin B
Target Apoptosis
Quantity 5 mg