Learn More
Methyl Benzethonium chloride, MP Biomedicals™
Product Code. 17881381 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17881381 10 g 10g
17891381 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17881381 Supplier MP Biomedicals Supplier No. 0219024010
  • ONLINE EXCLUSIVE PRICE
    5400.00 NOK
    5139.83 NOK / 10g
    Save 260.17 NOK
    4% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Chemical Identifiers
CAS 25155-18-4
Molecular Formula C28H44ClNO2
Molecular Weight (g/mol) 462.115
InChI Key QWZLBLDNRUUYQI-UHFFFAOYSA-M
PubChem CID 5702238
IUPAC Name benzyl-dimethyl-[2-[2-[2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC1=C(C=CC(=C1)C(C)(C)CC(C)(C)C)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]
Specifications
Color White
Quantity 10 g
Synonym methylbenzethonium chloride, methyl benzethonium chloride, unii-nn7590iuqx, nn7590iuqx, benzyldimethyl 2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium chloride, quat20, quat 20, prestwick_9, metilbenzetonio cloruro, benzyl-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium;chloride
Formula Weight 462.1
Physical Form Powder
Chemical Name or Material Methyl Benzethonium chloride
compliance-icons compliance-icons
Product Identifier
  • Methyl Benzethonium Chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methylbenzethonium chloride