Learn More
Methyl 3-bromo-2,4-dioxo-4-phenylbutyrate, 97%, Thermo Scientific™
Product Code. 15448805
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15448805
Brand: Thermo Scientific Alfa Aesar H34166.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59609-59-5
Molecular Formula C11H9BrO4
Molecular Weight (g/mol) 285.093
MDL Number MFCD00573907
InChI Key ZLWZFMIPUOKDJU-UHFFFAOYSA-N
Synonym methylbromodioxophenylbutanoate, methyl3-bromo-2,4-dioxo-4-phenylbutanoate, methyl 3-bromo-2,4-dioxo-4-phenylbutyrate, 3-bromo-2,4-dioxo-4-phenylbutanoic acid methyl ester, benzenebutanoic acid, b-bromo-a,g-dioxo-, methyl ester, benzenebutanoic acid,, ?-bromo-, a,y-dioxo-, methyl ester, a-bromo-, methyl 3-bromanyl-2,4-bis oxidanylidene-4-phenyl-butanoate
PubChem CID 2764337
IUPAC Name methyl 3-bromo-2,4-dioxo-4-phenylbutanoate
SMILES COC(=O)C(=O)C(C(=O)C1=CC=CC=C1)Br
Specifications
Melting Point 110°C to 112°C
Quantity 250 mg
Formula Weight 285.1
Percent Purity 97%
Chemical Name or Material Methyl 3-bromo-2,4-dioxo-4-phenylbutyrate