accessibility menu, dialog, popup

UserName

CAS RN 59609-59-5

H3COOOOBr

CAS RN 59609-59-5

IUPAC Name: methyl 3-bromo-2,4-dioxo-4-phenylbutanoate
Synonyms: methyl 3-bromo-2,4-dioxo-4-phenylbutanoate
Molecular Weight (g/mol): 285.09
Molecular Formula: C11H9BrO4
InChi Key: ZLWZFMIPUOKDJU-UHFFFAOYNA-N
SMILES: COC(=O)C(=O)C(Br)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results