missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(methylthio)propionate, 99%, Thermo Scientific™

Product Code. 11469447
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469447 5 g 5g
11479447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469447 Supplier Thermo Scientific Alfa Aesar Supplier No. B21979.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl 2-(methylthio)propionate is used in the contribution of overall aroma perception of the ripe fruit.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61366-76-5
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.193
MDL Number MFCD02683079
InChI Key QMPSBIWJVUCDKV-UHFFFAOYSA-N
Synonym methyl 2-methylthio propionate, methyl 2-methylsulfanyl propanoate, methyl 2-methylthio propanoate, dimethyl sulphoniopropionate, methyl 2-methylsulfanyl propanoate #, 2-methylthio propanoic acid methyl ester, 2-methylthiopropionic acid methyl ester, propanoic acid, 2-methylthio-, methyl ester
PubChem CID 548372
IUPAC Name methyl 2-methylsulfanylpropanoate
SMILES CC(C(=O)OC)SC

Specifications

Refractive Index 1.462
Quantity 5 g
Solubility Information Immiscible with water.
Formula Weight 134.2
Percent Purity 99%
Chemical Name or Material Methyl 2-(methylthio)propionate
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.