accessibility menu, dialog, popup

UserName

CAS RN 61366-76-5

IUPAC Name: methyl 2-(methylsulfanyl)propanoate
Synonyms: methyl 2-(methylsulfanyl)propanoate
Molecular Weight (g/mol): 134.19
Molecular Formula: C5H10O2S
InChi Key: QMPSBIWJVUCDKV-UHFFFAOYNA-N
SMILES: COC(=O)C(C)SC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results