missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chlorophenylacetate, 98%

Product Code. 10456052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10456052 5 g Glass bottle 5g
10512422 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10456052 Supplier Thermo Scientific Acros Supplier No. 351040050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57486-68-7 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57486-68-7
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00137445
InChI Key KHBWTRFWQROKJZ-UHFFFAOYSA-N
PubChem CID 93683
IUPAC Name methyl 2-(2-chlorophenyl)acetate
SMILES COC(=O)CC1=CC=CC=C1Cl

Specifications

Color Colorless
Density 1.2100g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5240 to 1.526
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.21
Linear Formula ClC6H4CH2CO2CH3
Beilstein 09, 448
Synonym methyl 2-chlorophenylacetate, methyl 2-2-chlorophenyl acetate, methyl 2-chlorophenyl acetate, 2-chlorophenyl acetic acid methyl ester, benzeneacetic acid, 2-chloro-, methyl ester, 2-chlorophenylacetic acid methylester, 2-chlorobenzeneacetic acid methyl ester, acmc-209lyj, methyl2-chlorophenylacetate
Formula Weight 184.62
Percent Purity 98%
Chemical Name or Material Methyl 2-chlorophenylacetate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-chlorophenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.