CAS RN 57486-68-7
CAS RN 57486-68-7
IUPAC Name:
methyl 2-(2-chlorophenyl)acetate
Synonyms:
methyl 2-(2-chlorophenyl)acetate
Molecular Weight (g/mol):
184.62
Molecular Formula:
C9H9ClO2
InChi Key:
KHBWTRFWQROKJZ-UHFFFAOYSA-N
SMILES:
COC(=O)CC1=CC=CC=C1Cl
1
–
1
of
1
results
1
Methyl 2-chlorophenylacetate, 98%
CAS: 57486-68-7 Molecular Formula: C9H9ClO2 Molecular Weight (g/mol): 184.62 MDL Number: MFCD00137445 InChI Key: KHBWTRFWQROKJZ-UHFFFAOYSA-N PubChem CID: 93683 IUPAC Name: methyl 2-(2-chlorophenyl)acetate SMILES: COC(=O)CC1=CC=CC=C1Cl