Learn More
Methyl 2-(3-oxetanylidene)acetate, 95%, Thermo Scientific™
Product Code. 15483858
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Product Code. 15483858
Brand: Thermo Scientific Alfa Aesar H57376.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1105665-34-6
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.127
MDL Number MFCD16140399
InChI Key QUBZUQPHJXYCSI-UHFFFAOYSA-N
PubChem CID 46839994
IUPAC Name methyl 2-(oxetan-3-ylidene)acetate
SMILES COC(=O)C=C1COC1
Specifications
Quantity 250 mg
Synonym methyl 2-oxetan-3-ylidene acetate, methyl oxetan-3-ylideneacetate, methyl 2-3-oxetanylidene acetate, methyl oxetan-3-ylidene acetate, methyl2-oxetan-3-ylidene acetate, methyl 2-oxetan-3-ylidene ethanoate, oxetan-3-ylidene-acetic acid methyl ester, oxetane-3-ylideneacetic acid methyl ester, 2-3-oxetanylidene acetic acid methyl ester
Formula Weight 128.13
Percent Purity 95%
Chemical Name or Material Methyl 2-(3-oxetanylidene)acetate