accessibility menu, dialog, popup

UserName

CAS RN 1105665-34-6

H3COOO

CAS RN 1105665-34-6

IUPAC Name: methyl 2-(oxetan-3-ylidene)acetate
Synonyms: methyl 2-(oxetan-3-ylidene)acetate
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: QUBZUQPHJXYCSI-UHFFFAOYSA-N
SMILES: COC(=O)C=C1COC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results