Learn More
Astell Scientific™ Nelldorn Glass Dryer
Product Code. 12753015
Change view
Click to view available options
Height (Metric):
430 mm
Width (Metric):
490 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Unit Size
12753015 430 mm 490 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12753015 Supplier Astell Scientific™ Supplier No. DCU012
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 07-01-2027
An economical and efficient method of drying and storing most containers, glassware and plasticware.
  • Unit can be used for most types of dispensing containers including plastics
  • Equipment can be left on the rack until drying is complete
  • Capable of holding up to 60 items with 15-20 minutes drying times
  • Wall mountable or free-standing
  • Thermostatically controlled
  • Economical, quick and efficient
  • Comes with a variable 120-minute timer
  • A large volume of air is drawn over the controlled heating elements and forced through each drying tube
  • Rapid flow of air dries glass or plastic quickly and without any damage to the apparatus
  • Very low power consumption
  • Stepped design allows the removal of any item without disturbing the remainder of the load and facilitates the run-off of water to the front of the unit where a small drain pipe prevents it collecting at the base
  • Air intake is recessed at the back of the unit to allow wall or bench mounting
  • Fitted with 60 rigid stain resistant nylon drying tubes with an adjustable timer which can be preset to switch off at your desired time
Specifications
Product Type Drying Rack
For Use With (Application) Can Hold 30 x 75 mm Tubes, 13 x 100 mm Tubes, 9 x 40 mm Tubes, and 8 x 152 mm Tubes
Includes Variable 120 minute timer
Height (Metric) 430 mm
Width (Metric) 490 mm
Voltage 220/240 V
Frequency 50 Hz