Learn More
Fisherbrand™ Meker Burner, Natural Gas
Product Code. 12882154 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Outer Inlet:
24 mm
30 mm
Height (Metric):
162 mm
192 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Inlet Height (Metric) Unit Size
12882154 30 mm 192 mm Each
12872154 24 mm 162 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12882154
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
Estimated Shipment: 26-02-2027
Add to basket
Add to basket
Fitted with a perforated steel head, which gives off multiple small but very hot flames.
  • The hottest flame temperature for all natural gas burners is between 900° and 1000°C at the tip of the blue flame cone
  • Constructed utilizing a heavy pressure diecast base painted in a blue chemically resistant acrylic finish
  • All burners tested for flame quality and leakage prior to packing
  • Tests are carried out at the standard gas pressure for each specific type gas
  • Temperatures dependent on mains pressure and the chemical construction of the gas which varies in different parts of the world
  • Standard mains pressure for natural gas is 20 millibars
  • Not fitted with an air regulator
Specifications
Product Type Meker Burner
Gas Type Natural Gas
Diameter (Metric) Outer Inlet 30 mm
Height (Metric) 192 mm
No. per Pack 1 Ea.