missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK6 Rabbit anti-Human, Mouse, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 5307.92 NOK
Specifications
| Antigen | MEK6 |
|---|---|
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite 555 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17781339
|
Proteintech
CL555-12745-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17669896
|
Proteintech
CL555-12745-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEK6 Polyclonal antibody apecifically detetects MEK6 antigen in Human, Mouse Samples. It is validated for Immunofluorescence.Specifications
| MEK6 | |
| Polyclonal | |
| Liquid | |
| Human, Mouse | |
| BC012009 | |
| MAP2K6 | |
| IgG | |
| Antigen affinity purification |
| Immunofluorescence | |
| CoraLite 555 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 5608 | |
| MEK6 fusion protein Ag3706 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title