missing translation for 'onlineSavingsMsg'
Learn More

MEK6 Rabbit anti-Human, Mouse, CoraLite 555, Polyclonal, Proteintech

Product Code. 17669896 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17669896 100 μL 100µL
17781339 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 17669896

Brand: Proteintech CL55512745100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEK6 Polyclonal antibody apecifically detetects MEK6 antigen in Human, Mouse Samples. It is validated for Immunofluorescence.

Specifications

Antigen MEK6
Applications Immunofluorescence
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate CoraLite 555
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC012009
Gene Symbols MAP2K6
Host Species Rabbit
Immunogen MEK6 fusion protein Ag3706
Purification Method Antigen affinity purification
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5608
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.