missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK6 Rabbit anti-Human, Mouse, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL555-12745-20UL
Additional Details : Weight : 0.00010kg
Description
MEK6 Polyclonal antibody apecifically detetects MEK6 antigen in Human, Mouse Samples. It is validated for Immunofluorescence.Specifications
MEK6 | |
Polyclonal | |
CoraLite 555 | |
BC012009 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human, Mouse | |
Liquid |
Immunofluorescence | |
1000 ug/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
MAP2K6 | |
MEK6 fusion protein Ag3706 | |
20 μL | |
5608 | |
Store at -20°C. Avoid exposure to light. | |
IgG |