Learn More
Fisherbrand™ Mega Rack™ Clinical Tube Rack
Shop All Fisherbrand Products
Click to view available options
Color:
Blue
White
Positions:
120
216
Holds:
Tubes (10 mL), Tubes (16 mm)
Tubes (13 mm), Tubes (5 to 7 mL)
Diameter (Metric) Opening:
13 mm
16 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Tube Rack |
| Array | 12 x 18 |
| Autoclavable | Autoclavable |
| Color | White |
| Holds | Tubes (13 mm), Tubes (5 to 7 mL) |
| Material | Polypropylene |
| Positions | 216 |
| Shape | Rectangular |
| Shelves | 3 Tiers |
| Stackable | Yes, When Empty |
| Show More |