Learn More
Thermographic Measurements Ltd Thermax Encapsulated Temperature Indicator
Product Code. 11750929
Click to view available options
Temperature Range (Metric):
110°C
121°C
127°C
138°C
154°C
160°C
166°C
171°C
177°C
182°C
188°C
204°C
216°C
224°C
232°C
241°C
254°C
40°C
49°C
54°C
60°C
65°C
71°C
77°C
88°C
93°C
missing translation for 'unitSize'
Pack of 50
Product Code. 11750929
missing translation for 'mfr': Thermographic Measurements Ltd M01ETHE127D01P1
Only null left
Add to basket
Add to basket
Single use, irreversible threshold indicator reports the highest temperature reached
  • Immediate Response
  • Permanent Record of Highest Temperature Achieved
  • Oil, Water, and Steam Resistant
  • Inexpensive and Easy to Use
  • Self-adhesive
  • Specifications
    Certifications/Compliance ISO 9001
    Disposable Single-use
    For Use With (Application) For laboratory research, medical services, instrumentation, electronics industry, automotive, aerospace, steel, metallurgy, plastics industry, food industry applications
    Height (English) 0.51 in.
    Height (Metric) 13 mm
    Humidity Range 40 to 60%
    Length (Metric) 12 mm
    Material Polyester Film, Acrylic Adhesive
    Measurement Time 2 to 5 seconds
    Model Thermax Encapsulated Temperature Indicator 127°C (261°F)
    Thermometer Fill Wax-melt Technology
    Weight (Metric) 9 g
    Product Type Irreversible Temperature Indicator
    Length (English) 0.47 in.
    Description Thermax Encapsulated Temperature Indicator 127°C (261°F))
    Max. Temperature (English) 261°F
    Max. Temperature (Metric) 127°C
    Min. Temperature (English) 261°F
    Min. Temperature (Metric) 127°C
    Temperature Range (English) 261°F
    Temperature Range (Metric) 127°C
    Calibrated Yes
    For Use With (Equipment) Electrical equipment, pumps, braking systems, water pumps, aeronautical equpment, military equipment
    Accuracy ±1.5°C
    Subdivision Single Temperature
    Scale Dual °C (°F)