Learn More
TMC Hallcrest™ Thermax Chillchecker Temperature Indicators
Product Code. 11737218
Change view
Click to view available options
Display:
-17°C
-2°C
-8°C
9°C
Unit Size:
Pack of 100
Product Code. Display unitSize
11737218 -2°C Pack of 100
11717218 -8°C Pack of 100
11767218 9°C Pack of 100
11707218 -17°C Pack of 100
4 options
Product Code. 11737218
Brand: TMC Hallcrest™ M01KTHEN02C01X1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provide an irreversible identification for temperature controled issues. TMC Hallcrest™ Thermax Chillchecker Temperature Indicators can be used for transporation and storage of goods applications.
  • Confirmation that the temperatures have not been exceeded.
  • Color changes to blue when temperature is reached.
  • Easy to use and read.
  • Self Adhesive.
  • Available temperatures between -17 to 20°C.
  • Accurate to +/-1°C
Specifications
Depth (English) 0.23 in.
Depth (Metric) 6 mm
Disposable Single-use
Material PVC
Width (English) 1.25 in.
Product Type Temperature Indicator
Accuracy ±1°C
Display -2°C
Width (Metric) 32 mm