missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEA1 Mouse anti-Human, Rat, Clone: 1H8C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66472-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
MEA1 gene ontology annotations related to this gene include spermatogenesis.Specifications
MEA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16626, Q5FVH7 | |
MEA1 | |
MEA1 Fusion Protein Ag0375 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Western Blot | |
1H8C2 | |
Unconjugated | |
MEA1 | |
HYS, male enhanced antigen 1, MEA, MEA 1, MEA1 | |
Mouse | |
Protein G | |
RUO | |
4201, 685131 | |
-20°C | |
Liquid |