missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEA1 Mouse anti-Human, Rat, Clone: 1H8C2, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | MEA1 |
---|---|
Clone | 1H8C2 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888375
|
Proteintech
66472-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16878375
|
Proteintech
66472-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MEA1 gene ontology annotations related to this gene include spermatogenesis.Specifications
MEA1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HYS, male enhanced antigen 1, MEA, MEA 1, MEA1 | |
MEA1 | |
IgG1 | |
Protein G | |
Antibody |
1H8C2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat, Human | |
Q16626, Q5FVH7 | |
4201, 685131 | |
MEA1 Fusion Protein Ag0375 | |
Primary | |
-20°C | |
MEA1 |