Learn More
MP Biomedicals Mccollum & Davis Salt Mixture No. 185
Product Code. 11485642 Shop All MP Biomedicals Products
Click to view available options
Quantity:
2 kg
Unit Size:
2kg
Product Code. 11485642
Brand: MP Biomedicals 0290284102
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Salt mixture containing calcium lactate and potassium phosphate.
Salt mixture containing calcium lactate and potassium phosphate.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Product Type Animal Diet
Disposable Single-use
Quantity 2 kg
Sterility Non-sterile
Storage Requirements Room Temperature
Length (English) 1 ft.
Length (Metric) 0.3048 m
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.